Learn More
HdmX/MDM4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300287AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Inhibits p53/TP53- and TP73/p73-mediated cell cycle arrest and apoptosis by binding its transcriptional activation domain. Inhibits degradation of MDM2. Can reverse MDM2-targeted degradation of TP53 while maintaining suppression of TP53 transactivation and apoptotic functions.Specifications
| HdmX/MDM4 | |
| Polyclonal | |
| Unconjugated | |
| MDM4 | |
| RP11-430C7.1, DKFZp781B1423, HDMX, MDMX, MGC132766, MRP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4194 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O15151 | |
| MDM4 | |
| Between 125 and 175 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.