Learn More
HdmX/MDM4 Recombinant Rabbit Monoclonal Antibody (BL-3-2F2), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$157.08 - $378.84
Specifications
Antigen | HdmX/MDM4 |
---|---|
Clone | BL-3-2F2 |
Applications | Immunoprecipitation, Western Blot |
Classification | Recombinant Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1256
![]() |
Bethyl Laboratories, Inc
A700000T |
20 μL |
Each for $157.08
|
|
|||||
50-157-1255
![]() |
Bethyl Laboratories, Inc
A700000 |
100 μL |
Each for $378.84
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Inhibits p53/TP53- and TP73/p73-mediated cell cycle arrest and apoptosis by binding its transcriptional activation domain. Inhibits degradation of MDM2. Can reverse MDM2-targeted degradation of TP53 while maintaining suppression of TP53 transactivation and apoptotic functions.Specifications
HdmX/MDM4 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O15151 | |
4194 | |
Between 125 and 175 | |
Primary | |
4° C | |
MDM4 |
BL-3-2F2 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
RP11-430C7.1, DKFZp781B1423, HDMX, MDMX, MGC132766, MRP1 | |
MDM4 | |
IgG | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.