Description
- Eliminate cleaning, prep and set-up costs with ready-to-use 20 L FISHERBRAND HDPE,Carboy.
- Supplied sterile for one-time use, the high-density polyethylene vessels are ideal for storing sterile fluids and pharmaceutical/biotech products.
- Features
- HDPE carboy with polypropylene closure
- Sterile
- Leakproof
Specifications
Specifications
Autoclavable | Not Autoclavable |
Capacity (Metric) | 20 L |
Color | Natural |
Disposable | Reusable |
Material | HDPE |
Mouth Style | Wide |
Product Type | Carboy |
Sterility | Non-sterile |