Learn More
Description
- Narrow mouth carboy with spigot
- Made from food grade resin HDPE
- HDPE closure with tag holes includes an o-ring for added security
- Spigot with gasket is leakproof and screws directly into a welded wall insert
- Heavy duty swing handle aids in carrying
- Resistant to many common chemicals
Specifications
Specifications
| Capacity (English) | 13 gal. |
| Capacity (Metric) | 50 L |
| Closure Material | HDPE |
| Color | White |
| For Use With (Application) | Ideal for storing and dispensing various laboratory solutions. Suitable for foodstuff. |
| Includes | Closure, 2 handles, Spigot |
| Material | HDPE |
| Mouth Style | Wide |
| Product Type | Carboy with Spigot |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
