Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDPTP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983764
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3764 Supplier Bioss Supplier No. BS15434R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plays a role in sorting of endocytic ubiquitinated cargos into multivesicular bodies (MVBs) via its interaction with the ESCRT-I complex (endosomal sorting complex required for transport I), and possibly also other ESCRT complexes. May act as a negative regulator of Ras-mediated mitogenic activity. Plays a role in ciliogenesis.

Specifications

Antigen HDPTP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTPN23
Gene Accession No. Q9H3S7
Gene Alias AI462446; HDPTP; HD-PTP; His domain-containing protein tyrosine phosphatase; KIAA1471; Protein tyrosine phosphatase TD14; protein tyrosine phosphatase, non-receptor type 23; Ptpn23; PTP-TD14; Tyrosine-protein phosphatase non-receptor type 23
Gene Symbols PTPN23
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HDPTP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25930
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.