Learn More
Invitrogen™ HDX Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113671
Description
Immunogen sequence: MNLRSVFTVE QQRILQRYYE NGMTNQSKNC FQLILQCAQE TKLDFSVVRT For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
HDX is the Highly Divergent Homebox, a protein coding gene, and has DNA binding function and RNA polymerase II transcription factor activity.
Specifications
| HDX | |
| Polyclonal | |
| Unconjugated | |
| HDX | |
| CXorf43; D030011N01Rik; Gm380; HDX; Highly divergent homeobox; RGD1563666 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 139324 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7Z353 | |
| HDX | |
| Synthetic peptide directed towards the N terminal region of human HDX (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.