Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Comprehensive Enteric Bacterial Control Panel (Inactivated Pellet)
Shop All Microbiologics Products

Click to view available options
Quantity:
6 Pellets
Description
Each pellet contains the microorganisms:
- Campylobacter jejuni
- Escherichia coli (two strains)
- Plesiomonas shigelloides
- Salmonella enterica serovar Typhimurium
- Shigella sonnei
- Vibrio parahaemolyticus
- Yersinia enterocolitica
Specifications
Specifications
| Biosafety Level | 1 |
| Content And Storage | 2 to 25°C |
| Format | Helix Elite |
| Manufacturing Origin | United States |
| Product Type | Helix Elite Control Reference Panels and Standards |
| For Use With (Application) | Molecular Testing |
| Packaging Type | Plastic Box |
| Quantity | 6 Pellets |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction