Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HE4/WFDC2 Rabbit anti-Human, Clone: 109, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29013150UL
Description
HE4/WFDC2 Monoclonal specifically detects HE4/WFDC2 in Human samples. It is validated for ELISA.Specifications
HE4/WFDC2 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
109 | |
ELISA 1:5000-1:10000 | |
dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5epididymal protein 4 | |
Recombinant Human HE4/WFDC2 Protein | |
50 μg | |
Cancer | |
10406 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction