Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104033 Supplier Invitrogen™ Supplier No. PA5104033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CLP1.

HEAB is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEAB
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLP1
Gene Accession No. Q5PQL4, Q92989, Q99LI9
Gene Alias AI462438; ATP/GTP-binding protein; cleavage and polyadenylation factor I subunit 1; CLP1; CLP1, cleavage and polyadenylation factor I subunit; CLP1, cleavage and polyadenylation factor I subunit, homolog; HAMAP-Rule:MF_03035}; hClp1; Heab; homolog of yeast CFIA subunit Clp1p; polyadenylation factor Clp1; polyadenylation factor Clp1 {ECO:0000255; Polynucleotide kinase Clp1; polynucleotide kinase Clp1 {ECO:0000255; polyribonucleotide 5'-hydroxyl-kinase Clp1; polyribonucleotide 5'-hydroxyl-kinase Clp1 {ECO:0000255; Pre-mRNA cleavage complex II protein Clp1; pre-mRNA cleavage complex II protein Clp1 {ECO:0000255; RGD1307679
Gene Symbols CLP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLP1(Accession Q92989), corresponding to amino acid residues Y313-H363.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10978, 311166, 98985
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.