Learn More
International Enviroguard™ Head and Face Hoods (Ninja Hoods)
Catalog No. p-7222260 Shop All International Enviroguard Systems Products
These Head and Face Hoods (Ninja Hoods) provide economical, lightweight protection against dirt, grime and particulates in non-hazardous environments.
These head and face covers (''Ninja Hoods'') offer dual protection - they protect your face from particulates while also protecting workplace products from contaminants you may generate such as saliva, hair, dander, and more. Wearing a head and face hood is a practical, win-win for you and your workplace. Better yet, this one product can replace separate items such as beard restraints and hair bouffant.
  • Lightweight polypropylene product that is an excellent one-piece replacement where separate beard restraints and hair bouffants are used
  • Common applications include food processing, food service, pharmaceuticals, cleanrooms and controlled environments, agriculture, healthcare, and more
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.