Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories Healgen™ External Controls: COVID-19/Flu A and B Ag Combo Rapid Test Cassette

Catalog No. 22026007
Change view
Click to view available options
For Use With (Equipment):
Laboratories
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-026-007 Laboratories
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-026-007 Supplier Germaine™ Laboratories Supplier No. GCFCPN2
Add to Cart
Add to Cart

Controls for lateral flow immunochromatographic assay intended for the qualitative detection and differentiation of influenza A, and influenza B nucleoprotein antigens and SARS-CoV-2 nucleocapsid antigens.

Lateral flow immunochromatographic assay intended for the qualitative detection and differentiation of influenza A, and influenza B nucleoprotein antigens and SARS-CoV-2 nucleocapsid antigen directly from anterior nasal swab specimens of individuals with signs and symptoms of respiratory infection consistent with COVID-19 by their healthcare provider within the first 5 days of symptom onset when tested at least twice over three days with at least 48 hours between tests. Clinical signs and symptoms of respiratory viral infection due to SARS-CoV-2 and influenza can be similar. Positive results do not rule out bacterial infection or co-infection with other viruses.

Detect Three Respiratory Viruses Simultaneously from a Single Sample: Faster diagnosis for improved patient care and cost-effectiveness

  • Rapid Detection: Results available in 15 minutes
  • Excellent Test Performance (comparator method RT-PCR)
  • Simple and Effective: CLIA Waived testing at the point-of-care for use with minimally trained healthcare professionals
  • External Quality Controls Available: Facilitates routine quality procedures
  • Shelf Life that Spans Viral Seasonality: 16 months from date of manufacture at room temperature storage

Specifications

Detectable Analytes Influenza A and Influenza B Nucleoprotein Antigens, SARS-CoV-2 Nucleocapsid Antigen
Format Test Cassette
For Use With (Equipment) Laboratories
Sample Type Nasal Swab
Type Nasal Swab Control
Disposable Single-use
Level 1, 2
Sterility Sterile
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.