Learn More
binx™ health io™ System
Catalog No. 22021835
Change view
Click to view available options
Sample Type:
Urine, Vaginal Swab
Catalog No. Sample Type
22-021-835 Urine, Vaginal Swab
1 options
This item is not returnable. View return policy
Order Info: Order eligibility is based on upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
Catalog No. 22-021-835 Supplier binx™ Supplier No. 3.001.001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Order Info: Order eligibility is based on upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
The system is a fully automated, rapid, FDA 510(K)-cleared, CLIA-waived instrument intended for use in point-of-care or clinical laboratory settings for Chlamydia trachomatis (CT) and Neisseria gonorrhoeae (NG) DNA detection by polymerase chain reaction (PCR) for both males and females.
The binx™ health io™ Instrument is the first ever 510(K), FDA-cleared, CLIA-waived, molecular point-of-care (POC) platform for chlamydia (CT) and gonorrhea (NG) detection in both males and females, enabling same-visit test and treatment.
  • The device is fully integrated and provides automated on-demand testing without the need for maintenance or calibration
  • Expands access to laboratory comparable testing results in near-patient settings such as primary care offices, urgent cares, student health centers, community clinics, emergency departments, and retail pharmacies
  • Provides the ability to test and treat within a single visit which offers improved patient outcomes such as reduced patient anxiety, lower transmission rates, fewer patients lost to follow-up, reduced empirical treatment, and less likelihood of complications or adverse events
  • It is an easy-to-use, desktop-sized instrument that can be operated by non-laboratory trained personnel in CLIA-waived settings
  • The device provides results comparable to a laboratory-based test for chlamydia and gonorrhea in about 30 minutes rather than days or weeks
Specifications
Type Accuracy Reader
Sample Type Urine, Vaginal Swab