Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

binx™ health io™ CT/NG Assay

Catalog No. 22021837
Change view
Click to view available options
Clia Complexity:
Waived
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22-021-837 Waived
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-021-837 Supplier binx™ Supplier No. 1.002.101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The CT/NG Assay is a single patient, single-use cartridge for the qualitative detection of chlamydia (CT) and gonorrhea (NG). The assay is performed on the binx™ health io™ Instrument.

The binx™ health io™ CT/NG Assay is intended for use with female vaginal swab specimens, collected either by a clinician or self-collected by a patient in a clinical setting; or male urine specimens, as an aid in the diagnosis of symptomatic or asymptomatic Chlamydia trachomatis and/or Neisseria gonorrhoeae infection.

For the detection of chlamydia (CT), the sensitivity and specificity rates were 96.1% and 99.1%, respectively, in female vaginal swab samples, whereas in male urine samples, the sensitivity and specificity rates were 92.5% and 99.3%, respectively. Conversely, for gonorrhea (NG), female vaginal samples exhibited sensitivity and specificity rates of 100.0% and 99.9%, while male urine samples demonstrated rates of 97.5% sensitivity and 100.0% specificity.

  • Fully automated, FDA-cleared, CLIA-waived, sample-in result-out assay performed in a self-contained cartridge utilizing ultra-rapid polymerase chain reaction (PCR) combined with proprietary electrochemical detection, enabling sensitivity and specificity equivalent to central lab performance
  • Provides the ability to test and treat within a single patient visit to offer improved patient outcomes such as reduced patient anxiety, lower transmission rates, fewer patients lost to follow-up, reduced empirical treatment, and less likelihood of complications or adverse events
  • It is an easy-to-use, desktop-sized instrument that can be operated by trained personnel in CLIA-waived settings

Specifications

Clia Complexity Waived
Detectable Analytes C. trachomatis, N. gonorrhoeae
Sample Type Male Urine, Vaginal Swab
Sensitivity High
Type Diagnosic Test
For Use With (Application) Chlamydia Testing
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.