Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PURELL™ Healthcare Advanced ULTRA NOURISHING™ Hand Sanitizer Foam

Shop All Gojo Industries, Inc. Products


Click to view available options
Volume (Metric):
1200 mL
Description
- Refill for PURELL™ ES4 Push-Style Hand Sanitizer Dispensers
- Unique blend of moisturizers and antioxidants, clinically proven to improve skin health in only 3 days
- Contains ingredients that help strengthen the skin's natural moisture barrier
- Reduces the appearance of redness and dryness in the skin
- Compatible with nitrile, vinyl, and latex gloves
- Hypoallergenic
- Dermatologist tested
- Contains vitamins B3 and E
- Chlorhexidine Gluconate (CHG) compatible formulation

Specifications
Specifications
Product Type | Hand Sanitizer |
Color | Clear |
Form | Foam |
Size | 1200 mL |
Volume (Metric) | 1200 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction