Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heartworm Antibody (HW1-39), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063168C
Description
Heartworm Monoclonal specifically detects Heartworm in Invertebrate samples. It is validated for Western Blot, ELISA.Specifications
Heartworm | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
NB100-63168 recognizes both male and female ether extracted parasite antigen of canine heartworm. Canine heartworm infection, caused by the filarial nematode Dirofilaria immitis is an insidious and potentially fatal condition transmitted by mosquitoes, which primarily affects the heart and lungs of dogs, but may also affect other species including cats, foxes, sea-lions, ferrets and humans. Canine heartworm disease, resulting ultimately in damage to the pulmonary vasculature system, is largely attributed to the immunological response of the host to the presence of the adult parasites, and in many cases their microscopic offspring (microfilaria) which circulate in the bloodstream, but growing evidence suggests the involvement of further factors/mediators. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
HW1-39 | |
Western Blot, ELISA | |
Dirofilaria immitus | |
Made to Invertebrates HEARTWORM | |
0.125 mL | |
Primary | |
Invertebrate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction