Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heartworm Chicken anti-Dog, HRP, Polyclonal , Abnova™

Catalog No. 89933717 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-717 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-717 Supplier Abnova Corporation Supplier No. PAB29674
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against dog Dirofilaria immitis.

Specifications

Antigen Heartworm
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Description Chicken polyclonal antibody raised against dog Dirofilaria immitis.
Dilution ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.5-5.0 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM HEPES (Stabilizer, 0.002% Thimerosal)
Host Species Chicken
Immunogen Dirofilaria immitis
Purification Method Antigen Affinity Purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Dog
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.