Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Fetal Bovine Serum, heat inactivated, qualified, Canada, One Shot™ format

Catalog No. p-7116269
Encompass_Preferred
Click to view available options
Quantity:
50 mL

Heat-inactivated serum in a One-Shot 50-mL bottle increases workflow benefits, while maintaining serum quality. The 50-mL bottle eliminates the need to aliquot and reduces the risk of contamination.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Serum category: Value (Standard)
    • For cell culture with robust cell lines
    • Good value for basic research
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Origin: countries meeting USDA importation requirements

Gibco sera: trusted quality for consistent performance

  • ISO 13485-certified, processed in FDA-registered facilities
  • Triple filtered at 0.1 μm

Heat-inactivated and convenient One-Shot 50-mL bottle format

Heat-inactivated serum in a One-Shot 50-mL bottle increases workflow benefits, while maintaining serum quality. The 50-mL bottle eliminates the need to aliquot and reduces the risk of contamination. Purchasing heat-inactivated serum eliminates the multiple steps necessary to heat inactivate and improves product integrity and consistency since the heat inactivation is done in a controlled environment.

One-Shot 50-mL bottle features include:

  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500-mL bottles
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Bovine
Age Fetal
Country of Origin Canada
Format One Shot
Product Type Fetal Bovine Serum
Quantity 50 mL
Serum Treatment Heat-inactivated
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Purity or Quality Grade Qualified
Product Line One Shot™
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.