Learn More
Thermo Scientific™ Heat Insulator (2 Blocks), EMD
Click to view available options
Dimensions (L x W x H):
107 x 143 x 80 mm
Description
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 years
Specifications
Specifications
Description | Heat Insulator for 2 block |
For Use With (Equipment) | Thermo Scientific™ Compact Dry Bath/Block Heater |
Type | Heat Insulator |
Dimensions (L x W x H) | 107 x 143 x 80 mm |