Learn More
Heidolph™ Heat-On™ Accessories
GREENER_CHOICESDP
Catalog No. p-9151001 Shop All Heidolph North America Products
Click to view available options
Type:
Adapter Plate
Safety Handle
For Use With (Equipment):
135 mm Hotplate
MR Hei Series Magnetic Stirrer
Accessories for Heat-On™ attachments.
  • Optional safety lifting handles have a simple and secure quick-release mechanism that allows the user to quickly attach the handles and remove the block from the heat source, even whilst it remains hot.
  • Aluminum adapter fits Heat-on™ blocks on 135 mm Hotplates (required for IKA and the Radleys Stirring Hotplate).
  • Specifications
    Description Heat-On Accessory
    For Use With (Equipment) MR Hei Series Magnetic Stirrer
    Type Safety Handle
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.