Learn More
GSC Go Science Crazy Heat Radiation Demonstration
SCI_ED
Catalog No. s13992 Shop All GSC Products
Encompass
Catalog No. S13992 Supplier GSC Go Science Crazy Supplier No. HEATRAD02
Only null left
Add to Cart
Add to Cart
Measure temperature against time and observe the effect of color on the rapidity of temperature increase. The GSC Heat Radiation Demonstration makes it easy to study what happens when heat reaches a destination.
  • This lesson looks at what happens when radiant heat reaches a destination
  • The apparatus is very simple, 6 plastic containers of various colors, each with a thermometer in it and a foam cover
  • A 25-watt incandescent light bulb provides the radiant heat source
  • In studying the impact of color on absorbing heat radiation you will measure temperature against time and observe the effect of color on the rapidity of temperature increase
Specifications
Product Type Demonstration
Includes Thermometer, foam cover, six plastic containers of various colors, and a 25-watt light bulb
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov