Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heat Transfer Kit
SDPSCI_ED

Catalog No. S53012
Click to view available options
Grade:
6 to 12

Observe and measure the transfer of heat between two water samples of different temperatures.

  • Includes two insulated containers with slotted lids, a U-shaped aluminum transfer bar and two thermometers
  • Grades 6 to 12

Specifications

Product Type Heat Transfer Kit
Grade 6 to 12
For Use With (Application) To observe and measure the transfer of heat between two water samples of different temperatures
Includes Two insulated containers with slotted lids, a u-shaped aluminum transfer bar and two thermometers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.