Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Heated Access Port
SDP

Catalog No. 08112029 Shop All BINDER Inc. Products
Click to view available options
Diameter (Metric):
150 mm

Accessory for Binder™ LIT MK Series.

  • 150 mm diameter
  • On left side
Includes:
  • Locking latch
  • Silicone plug

Specifications

Description Heated access port - On left side, including locking latch and silicone plug
For Use With (Equipment) BINDER Chambers Series LIT MK
Material Steel
Type Access Port
For Use With (Application) Battery Testing
Diameter (English) 5.9 in.
Diameter (Metric) 150 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.