Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Heated Product Shelf Chambers
SDP

Catalog No. 10400489 Shop All Labconco Corporation Products
Change view
Click to view available options
Plug Type:
British (UK)
China/Australia
India
North America
Saudi Arabia
Schuko
Height (Metric):
43.1 cm
62.4 cm
Includes:
3 shelves and clear chamber
5 shelves and clear chamber
Voltage:
115 V
230 V
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Height (Metric) Includes Voltage
10-400-489 North America 62.4 cm 5 shelves and clear chamber 230 V
10-400-481 North America 43.1 cm 3 shelves and clear chamber 115 V
10-400-482 North America 43.1 cm 3 shelves and clear chamber 230 V
10-400-483 Saudi Arabia 43.1 cm 3 shelves and clear chamber 230 V
10-400-484 Schuko 43.1 cm 3 shelves and clear chamber 230 V
10-400-485 China/Australia 43.1 cm 3 shelves and clear chamber 230 V
10-400-486 British (UK) 43.1 cm 3 shelves and clear chamber 230 V
10-400-488 North America 62.4 cm 5 shelves and clear chamber 115 V
10-400-487 India 43.1 cm 3 shelves and clear chamber 230 V
10-400-490 Saudi Arabia 62.4 cm 5 shelves and clear chamber 230 V
10-400-491 Schuko 62.4 cm 5 shelves and clear chamber 230 V
10-400-492 China/Australia 62.4 cm 5 shelves and clear chamber 230 V
10-400-493 British (UK) 62.4 cm 5 shelves and clear chamber 230 V
10-400-494 India 62.4 cm 5 shelves and clear chamber 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 10-400-489 Supplier Labconco™ Supplier No. 780801110
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Includes

5 shelves and clear chamber

Requires

FreeZone Freeze Dryer

Allows drying of bulk samples, or drying of serum bottles, vials or microplates. Labconco Heated Product Shelf Chambers are wireless and removable so that they may be placed in freezer for pre-freezing.

  • Short Chambers include three 10 in. diameter shelves and three temperature probes; Tall Chambers include five 10 in. diameter shelves and five temperature probes
  • Shelves are wireless and removable so that they may be placed in a freezer for pre-freezing
  • Each shelf is height adjustable to accommodate broad variety of sample sizes
  • Chamber connects to FreeZone Freeze Dryer for control and recording of shelf temperature from Lyo-Works display
  • Temperature may be set from −20° C to 60° C (−4° F to 140° F)
  • Each Chamber includes 12 in. diameter clear acrylic cylinder, vacuum release valve and 8 in. diameter opening to collector on bottom of chamber
  • Will fit 10.12 in. or 14.0 in. diameter condensers
  • For use with any FreeZone Freeze Dryer except Legacy models

Compliance

All models conform to UL Standard 61010-1 (60 Hz models), CAN/CSA C22.2 No. 61010-1 (60 Hz models), and CE Conformity marking (230V, 50 Hz models).

Warranty and Services

One year

Specifications

Type Heated Shelf
No. of Shelves 3
Temperature Range (Metric) -20°C to +60°C
Material Steel (Shelves), Acrylic (Chamber)
Certifications/Compliance UL, CAN/CSA, CE
Includes 5 shelves and clear chamber
Depth (English) 15.2 in.
Depth (Metric) 38.6 cm
Width (English) 15.2 in.
Width (Metric) 38.6 cm
Height (English) 24.6 in.
Height (Metric) 62.4 cm
Amperage 2A
Plug Type North America
Voltage 230 V
Frequency 50/60 Hz
Shipping Weight (English) 80 lb.
Shipping Weight (Metric) 36.29 kg
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.