Learn More
Labconco Heated Product Shelf Chambers with Valves
SDP
Heated Product Shelf Chambers with Valves for bulk drying and flask drying. Eight ports accommodate both 0.5 and 0.75 in. flask adapters. Includes 12 in. dia. acryl cylind and base.
Supplier:  Labconco 780802110
 View more versions of this product
Catalog No. 17-720-244
Only null left
Add to Cart
Request a Quote
Description
Description
  • Heated Product Shelf Chambers
  • Short (three shelves) or tall (five shelves) options
  • With valves
  • For bulk drying and flask drying
  • 10 in. dia. removable shelves
  • Temperature probes
  • Controlled heat settings
Specifications
Specifications
Heated Product Shelf Chamber
-20°C to +50°C
8
230 V
5
Acrylic
North America
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.