Learn More
Labconco Heated Product Shelf Chambers with Valves
Heated Product Shelf Chambers with Valves for bulk drying and flask drying. Eight ports accommodate both 0.5 and 0.75 in. flask adapters. Includes 12 in. dia. acryl cylind and base.
Supplier: Labconco 780802010
Description
- Heated Product Shelf Chambers
- Short (three shelves) or tall (five shelves) options
- With valves
- For bulk drying and flask drying
- 10 in. dia. removable shelves
- Temperature probes
- Controlled heat settings
Specifications
Heated Product Shelf Chamber | |
-20°C to +50°C | |
8 | |
230 V |
3 | |
Acrylic | |
North America |