Learn More
Restek™ Heater Block Retaining Nuts
SDP
Catalog No. 06611197 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890 GCs
Agilent 6850/6890 GCs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06-611-197 Agilent 5890 GCs
06-611-254 Agilent 6850/6890 GCs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-197 Supplier Restek™ Supplier No. 22080
Add to Cart
Add to Cart
For Agilent GC Split/Splitless Injectors
  • Aluminum construction
  • Meets or exceeds original manufacturer's performance
Specifications
Type Heater Block Retaining Nut
Material Aluminum
Quantity 1
For Use With (Equipment) Agilent 5890 GCs