Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEATR1/BAP28 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | HEATR1/BAP28 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-9486
![]() |
Bethyl Laboratories, Inc
A305082AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Involved in nucleolar processing of pre-18S ribosomal RNA. Involved in ribosome biosynthesis.Specifications
HEATR1/BAP28 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
BAP28, FLJ10359, protein BAP28, UTP10, UTP10, small subunit (SSU) processome component, homolog, UTP10, small subunit (SSU) processome component, homolog (yeast) | |
Between 1200 and 1250 | |
Primary | |
4° C | |
HEATR1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9H583 | |
217995, 55127 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title