Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEATR1/BAP28 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569487
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9487 Supplier Bethyl Laboratories, Inc Supplier No. A305083A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Involved in nucleolar processing of pre-18S ribosomal RNA. Involved in ribosome biosynthesis.

Specifications

Antigen HEATR1/BAP28
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene HEATR1
Gene Accession No. Q9H583
Gene Alias BAP28, FLJ10359, protein BAP28, UTP10, UTP10, small subunit (SSU) processome component, homolog, UTP10, small subunit (SSU) processome component, homolog (yeast)
Gene Symbols HEATR1
Host Species Rabbit
Immunogen Between 2094 and 2144
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55127
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.