Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEATR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575818 Supplier Invitrogen™ Supplier No. PA575818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Ribosome biogenesis factor. Involved in nucleolar processing of pre-18S ribosomal RNA. Required for optimal pre-ribosomal RNA transcription by RNA polymerase I. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEATR1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene HEATR1
Gene Accession No. Q9H583
Gene Alias AA517551; B130016L12Rik; BAP28; BC019693; FLJ10359; HEAT repeat containing 1; HEAT repeat-containing protein 1; HEAT repeat-containing protein 1, N-terminally processed; HEATR1; Protein BAP28; protein UTP10; RGD1564769; U3 small nucleolar RNA-associated protein 10 homolog; UTP10; UTP10, small subunit (SSU) processome component, homolog
Gene Symbols HEATR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human HEATR1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217995, 361262, 55127
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.