Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEATR2 Polyclonal Antibody

Catalog No. PIPA535890
missing translation for 'changeView'
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. PIPA535890 Supplier Invitrogen™ missing translation for 'supplierNo' PA535890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT-116. Predicted reactivity: Human (99%), Mouse (82%), Rat (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is essential for the preassembly or stability of axonemal dynein arms, and is found only in organisms with motile cilia and flagella. Mutations in this gene are associated with primary ciliary dyskinesia-18, a disorder characterized by abnormalities of motile cilia. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEATR2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene DNAAF5
Gene Accession No. Q86Y56
Gene Alias BC053401; C76907; CILD18; DNAAF5; dynein (axonemal) assembly factor 5; dynein assembly factor 5, axonemal; dynein axonemal assembly factor 5; dynein, axonemal, assembly factor 5; HEAT repeat containing 2; HEAT repeat-containing protein 2; HEATR2
Gene Symbols DNAAF5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HEATR2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54919
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.