Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEATR2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551229

Catalog No. PA551229


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total HEATR2 protein.

The protein encoded by this gene is essential for the preassembly or stability of axonemal dynein arms, and is found only in organisms with motile cilia and flagella. Mutations in this gene are associated with primary ciliary dyskinesia-18, a disorder characterized by abnormalities of motile cilia. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HEATR2
Polyclonal
Unconjugated
DNAAF5
BC053401; C76907; CILD18; DNAAF5; dynein (axonemal) assembly factor 5; dynein assembly factor 5, axonemal; dynein axonemal assembly factor 5; dynein, axonemal, assembly factor 5; HEAT repeat containing 2; HEAT repeat-containing protein 2; HEATR2
Rabbit
Antigen affinity chromatography
RUO
54919
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q86Y56
DNAAF5
Fusion protein of human DNAAF5.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.