Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEATR3 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318482UV
Description
HEATR3 Polyclonal antibody specifically detects HEATR3 in Human, Mouse samples. It is validated for Western BlotSpecifications
HEATR3 | |
Polyclonal | |
Western Blot | |
FLJ20718, HEAT repeat containing 3, HEAT repeat-containing protein 3 | |
Recognizes region between residues Between between 25 and 75.. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
55027 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction