Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEATR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559809
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559809 Supplier Invitrogen™ Supplier No. PA559809
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CFLLEVTTKD PSLVVAGEAL DALFDVFADG KEAERASIQI KLLSALKEFQ PVFKMKIRKE GRGNYSTDQL CVLDNVKMNL RRFIAYQETV EKRL Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

The protein encoded by this gene plays a role in ribosomal protein transport and in the assembly of the 5S ribonucleoprotein particle (5S RNP). The encoded protein also may be involved in NOD2-mediated NF-kappaB signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEATR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEATR3
Gene Accession No. Q7Z4Q2
Gene Alias C030036P15Rik; HEAT repeat containing 3; HEAT repeat-containing protein 3; HEATR3; RGD1308267; SYO1
Gene Symbols HEATR3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEATR3. Recombinant protein control fragment (Product #RP-98727).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55027
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.