Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HEATR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570172
Description
This target displays homology in the following species: Human: 100%.
HEATR4 is a protein coding gene.
Specifications
HEATR4 | |
Polyclonal | |
Unconjugated | |
HEATR4 | |
HEAT repeat containing 4; HEAT repeat-containing protein 4; HEATR4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
399671 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86WZ0 | |
HEATR4 | |
synthetic peptide directed towards the N terminal region of human HEATR4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction