Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEATR5A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983769
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3769 Supplier Bioss Supplier No. BS15440R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen HEATR5A
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HEATR5A
Gene Accession No. Q86XA9
Gene Alias C14orf125; HEAT repeat containing 5A; HEAT repeat-containing protein 5A; HEATR5A; KIAA1316
Gene Symbols HEATR5A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HEATR5A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25938
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.