Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEATR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556559 Supplier Invitrogen™ Supplier No. PA556559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSEDTIDFLE FKYCVSLRTQ ICQALIHLLS LASASDLPCM KETLELSGNM VQSYILQFLK SGAEGDDTGA PHSPQERDQM VRMALKHMGS IQAPTGDTAR RAIMGFLEEI LAVCFDSSGS QGAL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.

Amplification-dependent oncogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEATR6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEATR6
Gene Accession No. Q6AI08
Gene Alias 2700008B19Rik; 2700008B19Rik-like protein; ABC1; amplified in breast cancer 1; amplified in breast cancer protein 1; BB135312; BB249351; HEAT repeat containing 6; HEAT repeat-containing protein 6; HEATR6; RGD1566204
Gene Symbols HEATR6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEATR6. Recombinant protein control fragment (Product #RP-93170).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.