Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEATR7A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983771
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3771 Supplier Bioss Supplier No. BS15442R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen HEATR7A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MROH1
Gene Accession No. Q8NDA8
Gene Alias HEAT repeat containing 7A; HEAT repeat-containing protein 7A; HEATR7A; KIAA1833; maestro heat like repeat family member 1; maestro heat-like repeat family member 1; Maestro heat-like repeat-containing protein family member 1; MROH1
Gene Symbols MROH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HEATR7A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727957
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.