Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes
GREENER_CHOICE

Catalog No. p-7075460
Click to view available options
Diameter (Metric):
94 mm
100 mm
Height (Metric):
16 mm
20 mm
Sterility:
Non-sterile
Sterile

Used for easy stacking. Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes is designed for better heat resistance using hot agar.

  • Vented for better gas exchange
  • Heavy design for heat resistance using hot agar
  • Manufactured from crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Polystyrene
Shape Round
Sterility Non-sterile
Type Petri Dish
No. per Case 360
Vent Yes
Height (Metric) 20 mm
Diameter (Metric) 100 mm

For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.