Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ Heavy Duty 3607 CPE Shoe Covers
Shop All International Enviroguard Systems Products
Click to view available options
Color:
Blue
White
Description
CPE disposable shoe covers help protect shoes and surrounding environments from dust, dirt, grime, and liquids. More durable than polypropylene and often used in cleanrooms, hospitals, or industrial commercial sites. Help protect against chemicals, dirt, dust, liquid splash, non-hazardous particulates, and even shallow immersion
- Water resistant and come with a strong elastic closure
- One size fits most; fits up to a men's size 12 1/2 shoe
- Can be worn in indoor and outdoor environments
- Possible applications include healthcare, cleanrooms, inspections, movers, delivery services, food processing, contract work, and more
Specifications
Specifications
| Size | Universal |
| Material | CPE |
| Latex Free | Yes |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Standard |
| Packaging Quantity | 300/Cs. |
| Resistance | Dry Particulates, Splash |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction