Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ Heavy Duty 3407 CPE Shoe Covers
Shop All International Enviroguard Systems Products
Click to view available options
Size:
Universal
X-Large
Color:
Blue
White
Description
Disposable shoe covers are built to last even in the most rugged environments. Heavy-duty CPE shoe covers are made of coated polyethylene to help protect against non-hazardous particles, liquid splash, and shallow immersion. Diamond pattern bottom offers skid resistance, enhanced traction, and durability.
- Water resistant and come with a strong elastic closure
- One size fits most; Fits up to a men's size 12 1/2 shoe
- Elastic cuffs for a secure fit around the ankles
- Can be worn until damaged or contaminated
- Available in both White and Blue
- Possible applications include construction, healthcare, cleanrooms, food processing, contract work, pharmaceuticals, moving services, and more
Specifications
Specifications
Size | X-Large |
Material | CPE |
Latex Free | Yes |
Color | Blue |
Disposable | Single-use |
Sterility | Non-sterile |
Closure Type | Elastic Ankle |
Sole Type | Skid-resistant |
Seam Type | Molded |
Packaging Quantity | 300/Cs. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction