Learn More
United Scientific™ Heavy-Duty Epoxy Coated Wire Rack For Test Tubes
Shop All United Scientific Supplies, Inc Products

Description
- The racks are designed with 3 levels keeping the tubes secure, the rack bottom is slotted and helps center the tubes.
- These racks are offered in various configurations and in 2 set of colors, blue and teal.
- Durable epoxy coating
- Robust steel wire rack construction
- Autoclavable
- Available in multiple sizes and configurations
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Blue |
| Holds | Tubes (20 to 25 mm) |
| Material | Epoxy-coated Wire |
| Positions | 40 |
| Shape | Rectangular, Rounded Corners |
| Stackable | No |
| Sterility | Non-sterile |
| Product Type | Test Tube Rack |
| Shelves | None |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.