Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heavy Metals, plasma standard solution, Specpure™, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL

Catalog No. AA14657AP
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. AA14657AP

Supplier: Thermo Scientific Chemicals 014657AP

Only null left
Add to Cart

Heavy Metals, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heavy Metals, plasma standard solution
Name Note Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL
Physical Form Liquid
Concentration Matrix: 5% HNO3
MDL Number MFCD02091713
UN Number UN3264
Solubility Miscible with water
Quantity 500 mL
Grade Specpure

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.