Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563298 Supplier Invitrogen™ Supplier No. PA563298
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNDKGIGMGM TVPISFAVFP NEDGSLQKKL KVWFRIPNQF QSDPPAPSDK SVKIEERE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

The full-length protein encoded by this gene is an intracellular tetrapyrrole-binding protein. This protein includes a natural chemoattractant peptide of 21 amino acids at the N-terminus, which is a natural ligand for formyl peptide receptor-like receptor 2 (FPRL2) and promotes calcium mobilization and chemotaxis in monocytes and dendritic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEBP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEBP1
Gene Accession No. Q9NRV9
Gene Alias HBP; Hebp; Hebp1; heme binding protein 1; heme-binding protein 1; p22 HBP; p22HBP
Gene Symbols HEBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HEBP1. Recombinant protein control fragment (Product #RP-100445).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50865
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.