Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595780 Supplier Invitrogen™ Supplier No. PA595780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEPLQPDPG AAEDAAAQAV ETPGWKAPED AGPQPGSYEI RHYGPAKWVS TSVESMDWDS AIQTGFTKLN SYIQGKNEKE MKIKMTAPVT SYVEPGSGPF SESTITISLY IPSEQQFDPP RPLESDVFIE DRAEMTVFVR SFDGFSSAQK NQEQLLTLAS ILREDGKVFD EKVYYTAGYN SPVKLLNRNN EVWLIQKNEP TKENE; Positive Samples: COS-7, Mouse lung; Cellular Location: Cytoplasm, Mitochondrion.

Can promote mitochondrial permeability transition and facilitate necrotic cell death under different types of stress conditions. Does not bind hemin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEBP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Hebp2
Gene Accession No. Q9WU63, Q9Y5Z4
Gene Alias C6orf34; C6ORF34B; Hebp2; heme binding protein 2; heme-binding protein 2; Placental protein 23; PP23; Protein SOUL; putative heme-binding protein; SOUL
Gene Symbols Hebp2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-205 of human HEBP2 (NP_0551351).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23593, 308632, 56016
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.