Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HEC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578319
Description
Positive Control: HeLa, Blocking peptide (GTX70016-PEP) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
Specifications
HEC1 | |
Polyclonal | |
Unconjugated | |
NDC80 | |
2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10403, 301701, 67052 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with no preservative | |
O14777, Q9D0F1 | |
NDC80 | |
Synthetic peptide within amino acids 30-80 around Ser55. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction