Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111613
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111613 Supplier Invitrogen™ Supplier No. PA5111613
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CYESFMSGAD SFDEMNAELQ SKLKDLFNVD AFKLESLEAK NRALNEQIAR LEQEREKEPN RLESLRKLKA SLQGDVQKYQ Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 78%.

Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDC80
Gene Accession No. O14777
Gene Alias 2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3
Gene Symbols NDC80
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human HEC1. Recombinant protein control fragment (Product #RP-105391).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10403
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.