Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518089
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518089 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518089 Supplier Invitrogen™ Supplier No. PA518089
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NDC80
Gene Accession No. O14777
Gene Alias 2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3
Gene Symbols NDC80
Host Species Goat
Immunogen Synthetic peptide sequence (YEKKATLIKSSEE) corresponding to the C-terminus amino acids of NDC80 (aa 630-642).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10403
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.