Learn More
HEC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300771A
Description
The recommended shelf life for this product is 1 year from date of receipt.
HEC is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.Specifications
| HEC1 | |
| Polyclonal | |
| Unconjugated | |
| NDC80 | |
| HEC, HEC1, KNTC2, TID3, hsNDC80 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10403 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O14777 | |
| NDC80 | |
| Between 600 and C-term | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.