Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HECA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544632 Supplier Invitrogen™ Supplier No. PA544632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HKLNTFHVRM EDDAQVGQGE DLRKFILAAL SASHRNVVNC ALCHRALPVF Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

This gene encodes the homolog of the Drosophila headcase protein, a highly basic, cytoplasmic protein that regulates the re-entry of imaginal cells into the mitotic cycle during adult morphogenesis. In Drosophila, the encoded protein also inhibits terminal branching of neighboring cells during tracheal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HECA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HECA
Gene Accession No. Q9UBI9
Gene Alias dJ225E12.1; Gm869; H dJ225E12.1; HDC; hdc homolog, cell cycle regulator; HDCL; headcase homolog; headcase homolog (Drosophila); headcase protein homolog; HECA; hHDC; si:dkey-34f16.2
Gene Symbols HECA
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human HECA.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51696
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.