Learn More
Invitrogen™ HECA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140046
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RSSRYLGEFL KNAIHLEPHK KAMAGGHVFR NAHFDYSPAG LAVHRGGHFD TPVQFLRRLD LSELLTHIPR HKLNTFHVRM EDDAQVGQGE DLRKFILAAL SASHRNVVNC ALCHRALPVF EQFPLVDGTL FLSPSRHDEI EYDVP.
This gene encodes the homolog of the Drosophila headcase protein, a highly basic, cytoplasmic protein that regulates the re-entry of imaginal cells into the mitotic cycle during adult morphogenesis. In Drosophila, the encoded protein also inhibits terminal branching of neighboring cells during tracheal development.
Specifications
HECA | |
Polyclonal | |
Unconjugated | |
HECA | |
dJ225E12.1; Gm869; H dJ225E12.1; HDC; hdc homolog, cell cycle regulator; HDCL; headcase homolog; headcase homolog (Drosophila); headcase protein homolog; HECA; hHDC; si:dkey-34f16.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51696 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UBI9 | |
HECA | |
Recombinant protein corresponding to Human HECA. Recombinant protein control fragment (Product #RP-100957). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.